missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Thiamphenicol | |
166°C | |
C12H15Cl2NO5S | |
5 g | |
OTVAEFIXJLOWRX-NXEZZACHSA-N | |
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
27200 | |
356.21 | |
Crystalline Powder |
15318-45-3 | |
White | |
MFCD00467983 | |
thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O | |
356.214 | |
CHEBI:32215 | |
≥98.0% (HPLC,N) |
Chemical Identifiers
15318-45-3 | |
356.214 | |
OTVAEFIXJLOWRX-NXEZZACHSA-N | |
27200 | |
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
C12H15Cl2NO5S | |
MFCD00467983 | |
thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol | |
CHEBI:32215 | |
CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O |
Safety and Handling
EINECSNumber : (9)-0816
RTECSNumber : AB6680000
TSCA : No