missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiamine Hydrochloride (USP/FCC), Fisher Chemical

Manufacturer:  Fisher Chemical O4700100

Catalog No. O4700-100




Specifications

Specifications

≤0.025 AU (Solution) (USP)
1.4g/cm³
0.2% max. (FCC, USP)
Meets Requirements (USP)
White
100g
67-03-8
MFCD00012780
DPJRMOMPQZCRJU-UHFFFAOYSA-M
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
6202
337.28
98.0 to 102.0% (USP)
Thiamine Hydrochloride
Pass Test (FCC, USP)
Glass Bottle
≤5.0% (FCC, USP)
260°C
2.7 to 3.4 (USP)
C12H18Cl2N4OS
aneurine hydrochloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
337.263
CHEBI:49105
Solid
FCC/USP

Chemical Identifiers

67-03-8
337.263
DPJRMOMPQZCRJU-UHFFFAOYSA-M
6202
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
C12H18Cl2N4OS
MFCD00012780
aneurine hydrochloride
CHEBI:49105
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Safety and Handling

Safety and Handling

SDS Documents