Learn More
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
$125.53 - $729.45
Chemical Identifiers
CAS | 67-03-8 |
---|---|
Molecular Formula | C12H18Cl2N4OS |
Molecular Weight (g/mol) | 337.263 |
MDL Number | MFCD00012780 |
InChI Key | DPJRMOMPQZCRJU-UHFFFAOYSA-M |
Synonym | thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride |
PubChem CID | 6202 |
ChEBI | CHEBI:49105 |
IUPAC Name | 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride |
SMILES | CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1956022
|
Thermo Scientific Chemicals
A1956022 |
100 g |
Each for $125.53
|
|
|||||
AAA1956030
|
Thermo Scientific Chemicals
A1956030 |
250 g |
Each for $245.53
|
|
|||||
AAA195600B
|
Thermo Scientific Chemicals
A195600B |
1000 g |
Each for $729.45
|
|
|||||
Description
Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.
Solubility
Soluble in water, glycerol, ethanol, dimethyl sulfoxide and methanol. Insoluble in benzene, hexane, chloroform and ether.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
67-03-8 | |
337.263 | |
DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
6202 | |
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride |
C12H18Cl2N4OS | |
MFCD00012780 | |
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride | |
CHEBI:49105 | |
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] |
Specifications
67-03-8 | |
0.99 | |
MFCD00012780 | |
3851771 | |
14,9295 | |
Soluble in water,glycerol,ethanol,dimethyl sulfoxide and methanol. Insoluble in benzene,hexane,chloroform and ether. | |
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] | |
337.263 | |
CHEBI:49105 | |
99% | |
(dry wt.), may cont. up to 5% water |
∼250°C (decomposition) | |
C12H18Cl2N4OS | |
100 g | |
Hygroscopic; Light sensitive | |
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride | |
DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride | |
6202 | |
337.28 | |
Thiamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319-H500
EINECSNumber : 200-641-8
RTECSNumber : XI7350000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only