missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thexyltrichlorosilane, TCI America™

Manufacturer:  TCI America T25175G

 View more versions of this product

Catalog No. T25175G



Specifications

Specifications

Thexyltrichlorosilane
Yellow
5g
C6H13Cl3Si
2987
OMLLOLAFIFULFX-UHFFFAOYSA-N
trichloro(2,3-dimethylbutan-2-yl)silane
15211689
Clear Liquid at 20°C
187°C
16°C
18151-53-6
MFCD03412148
Trichloro(2,3-dimethyl-2-butyl)silane, Trichloro(1,1,2-trimethylpropyl)silane
CC(C)C(C)(C)[Si](Cl)(Cl)Cl
219.605
219.61
≥96.0% (GC)

Chemical Identifiers

18151-53-6
219.605
OMLLOLAFIFULFX-UHFFFAOYSA-N
15211689
CC(C)C(C)(C)[Si](Cl)(Cl)Cl
C6H13Cl3Si
MFCD03412148
Trichloro(2,3-dimethyl-2-butyl)silane, Trichloro(1,1,2-trimethylpropyl)silane
trichloro(2,3-dimethylbutan-2-yl)silane
Safety and Handling

Safety and Handling

TSCA : No

Documents