Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
Supplier:  Sonoco™ FPP31
 View more versions of this product
Catalog No. 03-530-012
Description
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
  • Provide refrigeration for transport of food, pharmaceuticals, and medical products.
  • Gel will phase change at 0° C.
  • PolarPack Foam Bricks maintain designated packing configurations throughout the trip.
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
  • Specifications
    Specifications
    18/Cs.
    9 in.
    22.8 cm
    Refrigerant Ice Pack
    31 oz.
    9 x 4 x 1.5 in. (22.8 x 10.16 x 3.81 cm)
    10.16 cm
    4 in.
    1.5 in.
    3.81 cm
    1.2 kg
    0°C (32°F)