Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
Manufacturer: sonoco™  TS356
 View more versions of this product
Catalog No. 03-527
  • / Case of 300 Each
Description & Specifications
Specifications
Material Polystyrene (PS) Foam
For Use With Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples
Quantity 1 Tubes
Capacity 3, 5, 7 or 10mL