missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe LabMailer Specimen Mailer
Lightweight, economical lab mailer shippers
Supplier: Sonoco™ 335
Description
- Hold screw-capped or rubber-capped tubes or vials and swabs
- Provide maximum breakage protection for known nonhazardous materials
- Protect frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use
Additional packaging may be required to conform to CFR72.2 specification (See Packaging Accessories Kit Mfr. No. 471).
Specifications
Lab Mailer for Vials, Tubes and Bottles | |
LabMailer | |
5.875 x 1.5 x 1.5 in. | |
Outside: 7 x 2.5 x 2.5 in.; Cavity: 5.875 x 1.5 x 1.5 in. |
Multi-tube, slide, and tube/slide combinations | |
7 x 2.5 x 2.5 in. | |
Holds specimen tubes, vials, swabs. Must order Model 336 for mailing sleeve. |