Sonoco™ ThermoSafe Insulated Storage and Transport Chests
Dry ice storage chests provide mobile, non-electrical low-temperature maintenance for a wide variety of laboratory and field applications. Ideal for dry ice or wet ice storage, hot storage, or field collection.
Manufacturer:  Sonoco™ TS302
 View more versions of this product
Catalog No. 11-676-11
Description
Description
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
  • Efficient, non deteriorating urethane foam insulation
  • Resist vapor, water, rust, corrosion
  • Washable
  • Interior corners are rounded for easy cleanup
  • Lid is hinged (except 50 lb. size) and includes support that keeps it upright
Order Info
Dollies not included
Specifications
Specifications
2 Spring Loaded Handles
Storage and Transport Chest
20.75 in.
23 in.
22 in.
Hinged
Ideal for dry ice and wet ice storage, short-term frozen and hot storage
Polyethylene
52.70 cm
58.42 cm
55.88 cm
23 x 22 x 20.75 in. (58.42 x 55.88 x 52.70 cm)
Sonoco™ ThermoSafe Aluminum Dollies (Fisher Cat. 11-676-24)
35 lb.
Documents
Accessories