Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe Insulated Shipper-PUR Containers: 3 in. Thick
Durable polyurethane-foam–molded shippers have twice the insulation value of EPS
Manufacturer: sonoco™  TSE90UPS
 View more versions of this product
Catalog No. 03-530-005
  • / Case of 2 Each
Description & Specifications
Specifications
Product Type Wall thickness: 3 in. (7.6cm)
Material Insulated Polyurethane (PUR) foam