Sonoco™ ThermoSafe™ GREENBOX™ Thermal Management Systems: Refrigerated (2-8°C)
GREENBOX is a sustainable packaging, designed for the safe transportation of temperature sensitive products.
$607.09 - $2,122.76
Specifications
Length (English) Exterior 12 in.
Width (English) Exterior 12 in.
Height (English) Exterior 12 in.
Product Type Temperature-controlled package shipper
Material Vacuum Insulated Panels (VIP)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Payload Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Payload Price Quantity  
03-600-128 Sonoco™
TS12UR1
8.31L Each for $1,102.68
03-600-130 Sonoco™
TS12UR3
5.44L Each for $1,193.85
03-600-131 Sonoco™
TS12UR4
5.21L Each for $1,367.00
03-600-129 Sonoco™
TS12UR2
5.16L Each for $1,150.74
03-600-126 Sonoco™
TS09UR1
Each for $607.09
03-600-127 Sonoco™
TS09UR2
Each for $768.15
03-600-132 Sonoco™
TS24UR3
18.63L Each for $2,122.76
Specifications
Description
Greenbox™ Reusable Insulated Shippers have been designed to utilize simple packaging configurations, reduce operational
complexity, and optimize total cost of ownership. GreenBoxs simple design and modular format increases reclamation and
refurbishment efficiencies. Ensure the efficacy of your valuable temperature-sensitive products and maintain regulatory
compliance throughout distribution with Greenbox.
  • ZERO Bench-Time™ refrigerant design means no waiting, greatly simplifies and accelerates the assembly process
  • Utilizes VIP insulation and PureTemp™ PCMs resulting in fewer components and lower weight, decreasing total cost of ownership
Description & Specifications
Specifications
12 in.
12 in.
Vacuum Insulated Panels (VIP)
12 in.
Temperature-controlled package shipper