Sonoco™ ThermoSafe Diagnostic Shipper, Three Tube Mailer System
Mailer secures up to three 10mL tubes
Supplier:  Sonoco™ 476
 View more versions of this product
Catalog No. 03-528-26
Description
Description
  • 95kPa certified receptacles (tubes or bags depending on kit)
  • Ready-to-use
  • Sufficient absorbent material to absorb maximum capacity of liquid
  • IATA Packing Instruction 650, Biological Substance, Category B label
Compliance
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples.
Specifications
Specifications
EPS 3-tube foam specimen mailer with lid
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Foam Mailer Only (no tubes)