Sonoco™ ThermoSafe Diagnostic Shipper, Eight Tube Mailing System
Leak-resistant, EPS foam mailer holds 30mL bottle, eight 10mL tubes, materials for insurance testing and other paperwork
$393.79 - $1,384.00
Specifications
Certifications/Compliance Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
For Use With Diagnostic materials
View More Specs
Includes: Complete set (Cat. No. 03-527C): 10mL vial tubes, foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with antitamper locking tabs and instructions. Cat. No. 03-527B includes eight-tube safety mailer only.
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Material Price Quantity  
Catalog Number Mfr. No. Includes Material Price Quantity  
03-527B Sonoco™
TS470
Safety Mailer (EPS Foam Unit Only) Expanded Polystyrene (EPS) Foam Case of 50 Each for $393.79
03-527C Sonoco™
TS472
10mL vial tubes, foam mailer with lid, absorbent material, water-resistant tape, pressure-lock plastic bag, 200 lb. test corrugated shipping carton with antitamper locking tabs and instructions Expanded Polystyrene (EPS) Foam and Cardboard, plastic tubes Case of 50 Each for $1,384.00
Specifications
Description
  • Tight-fitting plug cover accepts required absorbent material
  • Validated for the transport of diagnostic samples
Compliance
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
Meets U.S. Postal Regulations, DOT regulations, and IATA Packaging Instruction 650; tubes and shipping system validated for the transport of diagnostic samples
Diagnostic materials