Thermo Scientific™ ThermoFlex™ Recirculating Chillers
Optimized for the most demanding applications, high-performance ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain
Manufacturer:  Thermo Scientific™ 111101010000011
 View more versions of this product
Catalog No. 13-266-134
Specifications
Description
The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24000w/81964 BTU, is configurable to meet application demands.
Ease of Use
  • An intuitive user interface for ease of operation
  • Air and water filters that can be changed while unit is in operation
  • Innovative packaging for rapid installation
  • Quick start guide enables trouble-free system start-up
Superior Performance
  • Improved cooling capacity
  • Increased reliability
  • Ease of maintenance
Configurable Design
  • Wide range of available cooling capacities
  • Variety of available options
  • Installation flexibility
Recommended for:
  • Aerospace
  • Analytical
  • Biotech
  • Chemical
  • Industrial
  • Medical
  • Pharmaceutical
  • Research
  • University
Description & Specifications
Specifications
+/- 0.1°C
5 to 40°C
115V 50Hz
1.9 gal.
1400w
NRTL Certified to CSA Standards; CE marked
115V
10 to 40°C
1400w; Pumping Option PD1; 115V 60Hz
7.2L
1400w; Pumping Option PD1; 115V 60Hz
50Hz