Please login to your online account to display your discounted pricing
Thermo Scientific™ ThermoFlex™ Recirculating Chillers
Optimized for the most demanding applications, high-performance ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain
$6,721.44 - $8,205.67
Products
Catalog Number Mfr. No. Price Quantity    
13-266-131
thermo scientific™
101101110000001
Each for $7,460.42
13-266-130
thermo scientific™
101101010000006
Each for $6,721.44
13-266-134
thermo scientific™
111101010000011
Each for $7,466.69
13-266-135
thermo scientific™
111101110000008
Each for $8,205.67
Description & Specifications
The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24000w/81964 BTU, is configurable to meet application demands.
Ease of Use
  • An intuitive user interface for ease of operation
  • Air and water filters that can be changed while unit is in operation
  • Innovative packaging for rapid installation
  • Quick start guide enables trouble-free system start-up
Superior Performance
  • Improved cooling capacity
  • Increased reliability
  • Ease of maintenance
Configurable Design
  • Wide range of available cooling capacities
  • Variety of available options
  • Installation flexibility
Recommended for:
  • Aerospace
  • Analytical
  • Biotech
  • Chemical
  • Industrial
  • Medical
  • Pharmaceutical
  • Research
  • University