Please login to your online account to display your discounted pricing
Thermo Scientific™ Universal Platform Clamps for MaxQ™ 8000 Shakers
For use with MaxQ 8000 shakers
$231.42 - $266.96
Specifications
For Use With MaxQ 8000
Type Clamp
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) No. of Positions Price Quantity    
236028
thermo scientific™
236028
5L 6.0 Each for $257.98
236017
thermo scientific™
236017
4L 6.0 Each for $231.42
236018
thermo scientific™
236018
6L 4.0 Each for $266.96
Description & Specifications
Specifications
For Use With MaxQ 8000
Type Clamp
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Replacement screws also available separately.