Please login to your online account to display your discounted pricing
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns
Incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced bleed, and reduced susceptibility to oxidation
$661.78 - $1,782.46
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Price Quantity    
26-096-2760
thermo scientific™
260962760
0.15mm 0.15μm Each for $812.85
26-096-2750
thermo scientific™
260962750
0.15mm 0.15μm Each for $661.78
26-096-2940
thermo scientific™
260962940
0.15m 0.15μm Each for $1,782.46
Description & Specifications
  • Low polarity, silarylene phase with selectivity comparable to 5% diphenyl/95% dimethyl polysiloxane
  • Designed for low bleed and outstanding inertness
  • Similar to DB-5MS, VF-5ms, CP-Sil 8 Low-Bleed/MS, Rxi-5Sil MS, BPX5, ZB-5ms, Optima-5MS, SLB-5