Thermo Scientific™ TraceGOLD TG-5MS GC Columns
Widely used MS phase in gas chromatography
$670.26 - $1,805.32
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
26-098-2760 Thermo Scientific™
260982760
Each for $1,125.31
26-098-2750 Thermo Scientific™
260982750
Each for $670.26
26-098-2940 Thermo Scientific™
260982940
Each for $1,805.32
03-052-625 Thermo Fisher Scientific
260981180
Each for $703.88
Specifications
Description
  • Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Exceptional inertness ideal for analysis of active compounds
  • Equivalent to USP G27 phase
  • Similar to Rxi™-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8
Semivolatiles, Phenols, Amines, Residual solvents and solvent impurities, Drugs of abuse, Pesticides, PCB cogeners, Aroclor mixes