Please login to your online account to display your discounted pricing
Thermo Scientific™ TraceGOLD TG-5HT GC Columns
Offers extended operation up to 400°C, ideal for high temperature GC applications
$828.88 - $828.88
Catalog Number Mfr. No. Price Quantity    
thermo scientific™
Each for $828.88
Description & Specifications
  • Low polarity, 5% diphenyl/95% dimethyl polysiloxane
  • Lower bleed and better inertness than comparable high-temperature columns
  • Special design of fused silica tubing extends column lifetime by up to 40%
  • Similar to Rxi™-5HT, BD-5HT, VF-5HT, ZB-5HT
Phenols, Residual solvents, Drugs of abuse, Semivolatiles, Pesticides, PCBs, Solvent impurities