Please login to your online account to display your discounted pricing
Thermo Scientific™ TraceGOLD TG-1MS GC Columns
Exceptionally low bleed for optimal signal-to-noise ratio, sensitivity and mass spec integrity
$648.43 - $1,746.49
Specifications
Packing Material Silica
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Stationary Phase Length (Metric) Price Quantity    
26-099-2750
thermo scientific™
26099-2750
0.15mm TG-1MS 10m Each for $648.43
26-099-2760
thermo scientific™
26099-2760
0.15mm TG-1MS 20m Each for $796.45
26-099-2940
thermo scientific™
26099-2940
0.15mm TG-1MS 40m Each for $1,746.49
Description & Specifications
Specifications
Packing Material Silica
  • Non-polar, 100% dimethyl polysiloxane
  • Equivalent to USP G2
  • Similar to Rxi-1ms, DB-1, DB-1ms, HP-1, HP-1ms, Ultra-1, SPB-1, Equity-1, VF-1ms, CP-Sil 5 CB Low Bleed,/MS
Hydrocarbons, PCBs, Drugs of abuse, Gasoline range organics (GRO), Refinery gases, Essential oils, Pesticides