Please login to your online account to display your discounted pricing

Thermo Scientific™ TRACE™ TR-SimDist GC Columns

Feature a low polarity phase for high temperature analyses

$625.20 - $707.40

Specifications

Diameter (Metric) Inner 0.53mm
View More Specs
Products
Catalog Number Mfr. No. Stationary Phase Length (Metric) Price Quantity    

26-AS0-07P

 
thermo scientific™
26AS007P
TR-SimDist 5m Each for $640.83

26-AS0-25P

 
thermo fisher scientific
26AS025P
TR-SimDist 10m Each for $707.40

26-AS0-83P

 
thermo fisher scientific
26AS083P
TR-SimDist 5m Each for $625.20
Description & Specifications

Specifications

Diameter (Metric) Inner 0.53mm

  • Nonpolar phase, 100% Dimethyl polysiloxane
  • Optimized for simulated distillation analysis
  • High temperature limits
  • Strongly cross-linked
  • Similar to DB-HT Sim Dis, DB-2887, BPX1, Rtx-2887, HP-1, Petrocol 2887, Petrocol EX2887

High molecular weight hydrocarbons, Simulated Distillation, ASTM™ D2887, D6532