missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations

For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations

$350.23 - $350.23

Specifications

Certifications/Compliance DNase, RNase, pyrogen free
Autoclavable Yes
Color White
For Use With (Equipment) Tecan™ Genesis™, Freedom Evo™, Cavro™, patchXpress workstations
Material Polypropylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
21-402-416 Thermo Scientific™
MB902262XP
Nonsterile Case of 960 Each for $350.23
Specifications

Description

Clear pipet tips, Tecan style.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

DNase, RNase, pyrogen free
White
Polypropylene
50μL
Rack
Yes
Tecan™ Genesis™, Freedom Evo™, Cavro™, patchXpress workstations
Filtered Robotic Tip
96 Tips/Rack
Racked