Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Compact, lightweight benchtop furnaces
Manufacturer:  Thermo Scientific™ F4792580
 View more versions of this product
Catalog No. 10-505-60
Specifications
Description
Furnace, Muffle; Thermo Scientific Thermolyne; Heavy duty; 100 to 1200C; 1000w; Eight-step programmable control; 2L; 120V 50/60Hz; 8.3A
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
1000w
2L
100 to 1200°C
8.3A
50/60Hz
120V