Thermo Scientific™ Thermocouple for Portable Muffle Furnaces
For use with Thermolyne Benchtop Muffle Furnaces.
Manufacturer:  Thermo Scientific™ TC48X1A*
Catalog No. 10-553-10B
Description
Description
Thermocouple break protection cuts the power to the heating elements, preventing a thermocouple failure runaway condition.
Specifications
Specifications
Thermolyne Benchtop 1100°C Muffle Furnaces (10-552, 10-552-1)
Thermocouple