Thermo Scientific™ Support Frames for Heracell™ 150i CO2 Incubators
For use with Thermo Scientific Heracell™ 150i CO2 Incubators
$1,162.35 - $5,667.60
Specifications
Type Accessory, Support Frame
For Use With (Equipment) Heracell 150i CO2 incubator
View More Specs
Products 4
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
13-998-053
View Documents
Thermo Scientific™
50057161
7.3 in. 18.5 cm
Each for $1,162.35
13-998-054
View Documents
Thermo Scientific™
50051376
7.9 in. 20 cm
Each for $1,599.35
13-998-055
View Documents
Thermo Scientific™
50051436
30.7 in. 78 cm
Each for $1,843.48
13-998-056
View Documents
Thermo Scientific™
50056459
30.7 in. 78 cm
Each for $5,667.60
Description
Description
For use with Thermo Scientific Heracell™ 150i CO2 Incubators
  • Provide protection against floor contamination
  • Choice of three heights: 185mm (7.3 in.), 200mm (7.9 in.), or 780mm (30.7 in.)
  • Support frames can also be caster-mounted for easy maneuverability
Specifications
Specifications
Accessory, Support Frame
Heracell 150i CO2 incubator
SDS