Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
$173.98 - $1,571.91
Specifications
For Use With (Equipment) 1/8 in. cannula or probe
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
14-823-266
thermo scientific™
400101
Pack of 100 Each for $173.98
14-823-267
thermo scientific™
400102
Case of 1000 Each for $1,559.26
14-823-268
thermo scientific™
400104
Pack of 100 Each for $175.54
14-823-269
thermo scientific™
400105
Case of 1000 Each for $1,559.26
14-823-270
thermo scientific™
400107
Pack of 100 Each for $175.54
14-823-272
thermo scientific™
400109
Case of 1000 Each for $1,559.26
14-823-273
thermo scientific™
400110*
Pack of 100 Each for $175.54
14-823-271
thermo scientific™
400108
Case of 1000 Each for $1,571.91
14-823-274
thermo scientific™
400111
Case of 1000 Each for $1,559.26
Description & Specifications
Specifications
For Use With (Equipment) 1/8 in. cannula or probe
  • Full flow design removes small particulates
  • Fit securely on the end of a 1/8 in. cannula or sample probe
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.