Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ Phenolic Screw Thread Caps
Solid top with rubber liner
$145.17 - $164.54
Specifications
Color Black
Material Phenolic
Type Solid Top Screw Cap
Material Closure Phenolic
Material (Liner) PTFE/Rubber
View More Specs
Products
Catalog Number Mfr. No. GPI Thread Thread Size Price Quantity    
22-035114
thermo scientific™
200832
15-415 15-415 Pack of 125 Each for $164.54
14-824-537
thermo scientific™
200835
15-425 15-425 Case of 100 Each for $145.17
Description & Specifications
Specifications
Color Black
Material Phenolic
Type Solid Top Screw Cap
Material Closure Phenolic
Material (Liner) PTFE/Rubber