Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ Glass Dissolution Test Vessels
Precision formed with flat flange and hemispherical bottom
$207.98 - $455.43
Products
Catalog Number Mfr. No. Price Quantity    
14-823-121
thermo scientific™
100006
Each for $455.43
14-823-122
thermo scientific™
100008
Each for $366.80
22-037015
thermo scientific™
100002
Each for $207.98
Description & Specifications
  • Manufactured from Type 1, Class A borosilicate glass Precision formed with flat flange and hemispherical bottom Glass vessels are serialized Graduations in 50mL increments, 400mL to 1000mL
Meet all specifications of USP XXIV, Monograph 711
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.