Thermo Scientific™ SUN-SRi™ Dissolution Flasks
Calibrated “to contain” and “to deliver” with permanently etched graduation lines and a write-on patch
$280.26 - $338.72
Specifications
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
14-823-125 Thermo Scientific™
100014
Each for $280.26
14-823-124 Thermo Scientific™
100011*
Each for $338.72
Specifications
Description
  • Type 1, Class A borosilicate glass
  • Calibrated “to contain” and “to deliver” with permanently etched graduation lines and a write-on patch
  • Made specifically for the pharmaceutical industry
Compliance
Meets Federal Specifications NNN-F-289 and ASTM E288
Description & Specifications
Specifications
Clear