Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ Centrifugal Filter Devices
Combination filter/receiver tube unit for sample processing directly in centifuges
Manufacturer: thermo scientific™  62220NN
 View more versions of this product
Catalog No. 10-800-188
  • / Pack of 25 Each
Description & Specifications
Specifications
Max. Force 5,000xG
Housing Material Polypropylene
Membrane Material Nylon
Includes Filter housing; membrane; receiver tube
Volume (Metric) Up to 2mL
Pore Size 0.2μm
For Use With Benchtop and floor model centrifuges