Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ Centrifugal Filter Devices
Combination filter/receiver tube unit for sample processing directly in centifuges
Manufacturer: thermo scientific™  62215RC
 View more versions of this product
Catalog No. 10-800-186
  • / Pack of 100 Each
Description & Specifications
Specifications
Housing Material Polypropylene
Includes Filter housing; membrane; receiver tube
Max. Force 10,000xG
Volume (Metric) 200 to 750μL
For Use With Laboratory model microcentrifuges
Membrane Material Regenerated Cellulose
Pore Size 0.2μm