Please login to your online account to display your discounted pricing
Thermo Scientific™ SUN-SRi™ Centrifugal Filter Devices
Combination filter/receiver tube unit for sample processing directly in centifuges
Manufacturer: thermo scientific™  64520NN
 View more versions of this product
Catalog No. 10-800-198
  • / Pack of 25 Each
Description & Specifications
Specifications
Housing Material Polypropylene
Includes Filter housing; membrane; receiver tube
Max. Force 5,000xG
Volume (Metric) Up to 2mL
For Use With Benchtop and floor model centrifuges
Membrane Material Nylon
Pore Size 0.45μm