Thermo Scientific™ SUN-SRi™ Calibrated To Deliver Non-Standard Size Volumetric Pipets
Manufactured from Type 1, Class A borosilicate glass
Supplier:  Thermo Scientific™ 205640
 View more versions of this product
Catalog No. 14-823-245
Description
Description
All pipets have standard size tip openings.
Specifications
Specifications
To Deliver
A
0.422 oz.
Volumetric Pipet
ASTM E969
Borosilicate Glass
12.5 mL