missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SUN-SRi™ 8mm Microsampling Vials

Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.

Manufacturer:  Thermo Scientific™ 200034

 View more versions of this product

Catalog No. 14-823-134



Specifications

Specifications

8mm
1.25 in.
100 Pack
Round bottom
<5μL
6 mm
300 μL
Crimp
32mm
Round
Microsampling Vial
Clear
Borosilicate Glass
Documents