Thermo Scientific™ SUN-SRi™ 11mm Snap Caps
Easy to apply caps eliminate crimping and decrimping of closures.
Manufacturer:  Thermo Scientific™ 501385
 View more versions of this product
Catalog No. 14-823-399
Description
Description
  • Preassembled caps with septa for use with StepVial System Snap/Crimp Vials
  • Effective evaporation resistance—apply with simple thumb pressure
  • Audible click ensures a secure seal has been applied
Snap caps are not compatible with a standard 11mm aluminum seal finish.
Specifications
Specifications
11mm
Snap Cap
No
PTFE/Silicone
Orange
Documents