missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SUN-SRi™ 11mm Aluminum Seals

For StepVials™, wide opening and standard opening 12×32mm aluminum seal vials

Manufacturer:  Thermo Scientific™ 501310

 View more versions of this product

Catalog No. 14-823-377



Description

Description

  • Pre-assembled seals and septa

Specifications

Specifications

Silver
PTFE/Silicone for GC
Aluminum
1000 Pack
No
11 mm
Crimp Cap
SDS
Documents