missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SUN-SRi™ 10-425 Wide Opening Vials

Provide increased target area for easier sample preparation

Manufacturer:  Thermo Scientific™ 501353

 View more versions of this product

Catalog No. 14-823-391



Requires : Requires the use of 10-425 screw thread closures and 10mm septa

Description

Description

  • Manufactured of Clear, Type 1 Class A or Amber, Type 1 Class B borosilicate glass

Specifications

Specifications

10-425
0.23 in.
1.25 in.
Wide
Vial
12 mm
750 μL
Screw
6mm
32mm
100 Pack
<14μL
Polypropylene
Documents