Thermo Scientific™ Spectronic™ Disposable IR Cards for IR100 FT-IR Spectrometer
$286.64 - $327.36
Specifications
Type IR Card
For Use With (Application) For qualitative IR transmission analysis of organic liquids, materials soluble in organic solvents, semisolids, and pastes and quick analysis of many sample types
For Use With (Equipment) For IR100 FT-IR spectrometer
Includes Two sampling areas
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
14-200-18 Thermo Scientific™
0020300
Polyethylene Pack of 25 Each for $327.36
14-200-19 Thermo Scientific™
0020301
PTFE Pack of 25 Each for $286.64
Specifications
Description
  • For quick analysis of many sample types
  • Polyethylene or polytetrafluoroethylene
  • Two sampling areas
Description & Specifications
Specifications
IR Card
For qualitative IR transmission analysis of organic liquids, materials soluble in organic solvents, semisolids, and pastes and quick analysis of many sample types
For IR100 FT-IR spectrometer
Two sampling areas