Please login to your online account to display your discounted pricing
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer
The new standard for rugged, simple-to-use visible spectrophotometers for teaching and routine applications.
$2,939.77 - $2,939.77
Products
Catalog Number Mfr. No. Price Quantity    
S06240
thermo scientific™
840282500
Each for $2,939.77
Description & Specifications
Delivering quick, convenient measurements in the hands of novice users, the SPECTRONIC 200 spectrophotometer is the perfect fit for the teaching lab.
  • Modern on-board software provides total control without a computer
  • Coarse/fine wavelength setting
  • Adjustable tilt screen
  • Sealed keys
  • Classroom-friendly, adaptable sample compartment
  • Convenient cuvet rack compartments
  • Sloping surfaces discourage balancing of samples on unit