Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Legend™ T3/RT3 Swinging Bucket Rotor
Versatile high-throughput rotor for use with Legend™ T3 and RT3 centrifuges
$3,219.81 - $5,612.70
Specifications
Includes Rotor body only
View More Specs
Includes:  Rotor body only
Requires:  Buckets or carriers (order separately)
Products
Catalog Number Mfr. No. Max. RCF Speed Price Quantity    
05-376-293
thermo scientific™
11210823
Pack of 4 Each for $4,227.70
05-376-261
thermo scientific™
11174154
3,398xG 4,000rpm Pack of 4 Each for $3,219.81
05-376-292
thermo scientific™
11174223
3094xG 4,000rpm Pack of 4 Each for $5,612.70
Description & Specifications
Specifications
Includes Rotor body only
Accepts buckets or carriers to spin tubes, bottles, microplates and special containers.
  • Capacity: 4 × 750mL
  • Designed to be loaded into centrifuge with buckets/carriers already in place
  • Sealed buckets 05-376-262 offer biohazard protection
  • Transparent sealing caps allow visual detection of biological risks
Tube adapters sold separately
Sealed bucket 05-376-262 conforms to CAMR,™ Porton Down (UK) standards for biological containment.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.