Please login to your online account to display your discounted pricing

Thermo Scientific™ Sorvall™ Legend™ T3/RT3 Swinging Bucket Rotor

Versatile high-throughput rotor for use with Legend™ T3 and RT3 centrifuges

Manufacturer: thermo scientific™  11174218

 View all options for this product

Buckets or carriers (order separately)

Catalog No. 05-376-262


Description & Specifications


Type Sealed bucket
Includes Rotor body only
Capacity (Metric) 750mL
Tube Size 95mm
Speed 4,000rpm
For Use With (Equipment) CL40 and FL40 centrifuge and M4 High throughput swing out rotor
Max. RCF 3,398xG

Accepts buckets or carriers to spin tubes, bottles, microplates and special containers.

  • Capacity: 4 × 750mL
  • Designed to be loaded into centrifuge with buckets/carriers already in place
  • Sealed buckets 05-376-262 offer biohazard protection
  • Transparent sealing caps allow visual detection of biological risks