Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Swinging Bucket Rotors
For use with Legend T1/RT1 centrifuges
Manufacturer: thermo scientific™  11175612
 View all options for this product
Buckets (order separately)
Catalog No. 05-376-106
Description & Specifications
Type Sealing lid
Includes Rotor body only
Capacity (Metric) 200mL
Accept buckets to spin a wide range of tube styles, including standard, conical, skirted and Vacutainers.
  • Two capacities: 4 × 200mL and 4 × 280mL
  • Sealing lids 05376106 and 05376126 provide biocontainment protection per CAMR (HPA) Porton Down, UK