Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Swinging Bucket Rotors
For use with Legend T1/RT1 centrifuges
$552.68 - $3,611.34
Specifications
Speed 4,100rpm
View More Specs
Includes: Rotor body only
Requires: Buckets (order separately)
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) For Use With (Equipment) Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) For Use With (Equipment) Type Price Quantity  
05-376-104 Thermo Scientific™
11210246
4 x 200mL S41 swing bucket Each for $1,922.81
05-376-105 Thermo Scientific™
11175611
200mL S41 swing out rotor Bucket Pack of 4 Each for $1,902.26
05-376-125 Thermo Scientific™
11210436
280mL T41 swing out rotor for Thermo Scientific™ IEC™ CL31 centrifuge Standard bucket Pack of 4 Each for $3,611.34
05-376-126 Thermo Scientific™
11175712
280mL T41 swing out rotor Sealing lid Pack of 4 Each for $552.68
05-376-127 Thermo Scientific™
11175752
4 x 50mL Thermo Scientific™ IEC™ T41 swing out rotor Bucket Pack of 4 Each for $2,133.98
05-376-129 Thermo Sci Centrifu Bench
11175763
15 x 5/7mL Thermo Scientific™ IEC™ T41 swing out rotor Bucket Pack of 4 Each for $2,121.11
Specifications
Description
Accept buckets to spin a wide range of tube styles, including standard, conical, skirted and Vacutainers.
  • Two capacities: 4 × 200mL and 4 × 280mL
  • Sealing lids 05376106 and 05376126 provide biocontainment protection per CAMR (HPA) Porton Down, UK
Order Info
Tube adapters sold separately.
Compliance
Sealed bucket 05376262 conforms to CAMR,™ Porton Down (UK) standards for biological containment.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
4,100rpm