Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Swinging Bucket Rotors
For use with Legend T1/RT1 centrifuges
Manufacturer: thermo scientific™  11175752
 View more versions of this product
Buckets (order separately)
Catalog No. 05-376-127
  • / Pack of 4 Each
Description & Specifications
Specifications
Type Bucket
Capacity (Metric) 4 x 50mL
Speed 4,100rpm
Includes Rotor body only