Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Swinging Bucket Rotors
For use with Legend T1/RT1 centrifuges
Manufacturer:  Thermo Scientific™ 11210436
 View more versions of this product
Catalog No. 05-376-125
Requires : Buckets (order separately)
Specifications Description & Specifications
Specifications
Standard bucket
280mL
4,100rpm
3,082xG
Rotor body only
77mm
T41 swing out rotor for Thermo Scientific™ IEC™ CL31 centrifuge