Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Legend™ T1/RT1 Fixed-Angle Rotors
For use with Legend T1/RT1 centrifuges
$1,989.15 - $4,745.17
Products
Catalog Number Mfr. No. Price Quantity    
05-376-176
thermo scientific™
11210437
Each for $4,745.17
05-376-187
thermo scientific™
11175741
Each for $1,989.15
Description & Specifications
  • Carbon fiber rotor 05376789 is lightweight for easier handling and has two rows for higher-volume microliter sample throughput
  • Sealed microtube rotor 05376187 provides biocontainment protection per CAMR (HPA) Porton Down, UK
Tube adapters sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.