Please login to your online account to display your discounted pricing
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Rectangular Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
$238.00 - $330.81
Specifications
For Use With (Equipment) 4-place Swing rotor with rectangle bucket
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
75002021
thermo scientific™
75002021
Each for $330.81
13-100-603
thermo scientific™
75005386
Each for $238.00
Description & Specifications
Specifications
For Use With (Equipment) 4-place Swing rotor with rectangle bucket
Adapters sold individually; order correct quantity to equip rotor.