missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Shandon™ Pathcentre™ Tissue Processor Accessories
For use with the Epredia™ Shandon™ Tissue Processor
$53.51 - $2,491.83
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
75210161
|
Epredia™
75210161 |
Wax Disposal Bag | 10 Pack |
Case of 10 Each for $862.87
|
|
28-617-65
|
Epredia™
A75210193 |
Reagent Storage Bottle | 3 Pack |
Case of 3 Each for $2,491.83
|
|
28-609-10
|
Epredia™
66110048 |
Airbag | 4 Pack |
Case for $53.51
|
|
Specifications
Tissue processor |