missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Immu-Mount™
Non-permanent mountant for use with frozen sectioning, immunoperoxidase techniques and immunofluorescent applications.
Supplier: Epredia™ 9990414
Description
- Economical, organic, overnight-drying liquid coverslip
- Aqueous base for immunostaining and other techniques
- Non-fluorescing medium with a low viscosity
- Resists fading and polymerizes quickly for easier handling of finished slides
- Refractive index: 1.495 ±0.002 (liquid); 1.586 ± 0.002 (dry)
- Viscosity: Low (120-160 cps at 24°C)
- pH: 8.0 from 8.8 at 21 through 24°C
Specifications
For use with frozen section, immunoperoxidase techniques, and immunofluorescent preparations | |
Shandon Immu-mount | |
1 L |