Thermo Scientific™ Shandon™ Humidity Chamber
Ideal for most immunostaining procedures as well as other laboratory techniques
Manufacturer:  Thermo Scientific™ SH198
 View more versions of this product
Catalog No. 28-610-22
Includes: Two 24-slide acrylic tray spacers with legs (keeps aluminum slide trays separated); Two 20-slide aluminum slide trays
Description
Description
Maintains moist environment during incubation steps.
  • Heavy, flanged cover locks in moisture
  • Durable, clear acrylic chamber will accommodate three aluminum slide trays
  • Acrylic slide tray spacers and aluminum trays also available separately
Order Info
Chamber dimensions D x W x H: 14.12 x 8 x 3 in. (35.9 x 20.3 x 7.6cm)
Specifications
Specifications
Humidity chamber
Acrylic
Documents