Thermo Scientific™ Shandon™ Acrylic Tray Spacer
Ideal for most immunostaining procedures as well as other laboratory techniques
Manufacturer:  Thermo Scientific™ 198
Catalog No. 198
Includes: Two 24-slide acrylic tray spacers with legs (keeps aluminum slide trays separated); Two 20-slide aluminum slide trays
Description
Description
Maintains moist environment during incubation steps.
  • Heavy, flanged cover locks in moisture
  • Durable, clear acrylic chamber will accommodate three aluminum slide trays
  • Acrylic slide tray spacers and aluminum trays also available separately
Order Info
Chamber dimensions D x W x H: 14.12 x 8 x 3 in. (35.9 x 20.3 x 7.6cm)
Specifications
Specifications
Healthcare
Acrylic Tray Spacer
Acrylic
Humidity chamber
Documents